Learn More
DWK Life Sciences Wheaton™ Stopper Leash For B.O.D. Bottles
Ensures stoppers do not become lost when collecting field samples
Brand:  DWK Life Sciences 776580
Product Code. 12477171
Description
Description
  • 10mm (0.4 in.) holes at either end expand to fit securely around both stoppers and bottle necks
  • Keeps stopper secured to the neck of the bottle
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stoppers and bottle necks
Rubber