Description
- Designed for optimum freedom of movement, the B0610 has a short wrist cuff.
- The uncoated, seamless nylon liner reduces irritation and allergy risks while maintaining dexterity and tactility.
Specifications
Specifications
Coating Material | Uncoated |
Color | White |
Disposable | Reusable |
Hand | Ambidextrous |
Laboratory/Cleanroom Use | Yes |
Latex Free | Yes |
Material | Nylon |
Product Type | Glove |
Size | 6 |
Sterility | Non-sterile |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Product Title
SHOWA™ White General Purpose Gloves, Model B0610 >