Learn More
Fisherbrand™ White Polypropylene Caps Product Code.: 16134597
Add to basket
Fisherbrand™ White Polypropylene Caps
Product Code. 16134597 Shop All Fisherbrand Products
18-400, PTFE/Polyethylene Foam, 100
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
83-400
89-400
Closure Liner:
PTFE/Polyethylene Foam
Polyethylene Foam
Pulp/Vinyl
Quantity:
100
144
288
370
500
1300
2100
Product Code. 16134597
Add to basket
Add to basket
White polypropylene screw caps provides clear surface for writing on cap
  • Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
  • PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
  • Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
Color White
Quantity 100
Closure Color White
Neck Finish 18-400
Closure Included No
Product Type Lined Cap
Closure Liner PTFE/Polyethylene Foam
Closure Material Polypropylene
Product Content Correction
Product Title
Fisherbrand™ White Polypropylene Caps >