Learn More
BRAND™ Wide Neck Erlenmeyer Flask
Wide Neck Erlenmeyer Flask features a beaded rim and graduation marks.
Brand: BRAND™ 92814
100.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Borosilicate Glass 3.3
- DIN ISO 24450
- With beaded rim and graduation
Specifications
25 mL | |
Wide | |
Beaded |
Borosilicate Glass (3.3) | |
Yes |
Special Offers
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
Product Content Correction
Product Title
BRAND™ Wide Neck Erlenmeyer Flask > Capacity: 25 mL