Learn More
Bel-Art™ Wilmad™ O-Ring for Spinner
O-Ring for spinner is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
Brand: Bel-Art™ 66000-0057
This item is not returnable.
View return policy
Description
- Low cost alternative to the full VT version for sample changers.
- Weight and susceptibility matched to originals.
- Can be mixed with originals in sample changer operation.
- VT operation of -150°C to 200°C
- Does not jam at top of upper barrel during insert operation.
- Tachometer strip now with foil protection for improved life span.
- Weight compatible with room temperature spinners.
- Very cost effective as compared to the originals.
Specifications
Agilent/Varian NMR Spectrometer | |
Spinner Turbine | |
V-GFK-10 |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Bel-Art™ Wilmad™ O-Ring for Spinner > For Use With: 10 mm NMR Tubes