Learn More
Cofra Women's Safety Shoes, Paula Series
Women's Safety Shoes have breathable leather uppers for comfort, steel toecaps for extra protection and tough anti-static eva-nitrile foodbeds.
Brand: Cofra 76590-000_36
This item is not returnable.
View return policy
Description
- Lace-up general purpose safety shoes
- Water-repellant breathable black nubuck uppers
- Steel toecaps resistant to 200J
- Tough EVA-nitrile anti-static footbed
- Breathable polyamide internal lining
- Slip-resistant polyurethane soles
Specifications
Lace-up | |
EVA-Nitrile | |
Leather | |
Steel Toe | |
Anti-slip | |
Steel | |
Women's |
Black | |
Polyamide | |
Polyurethane | |
3 | |
Polyurethane | |
EN ISO 20345:2011 | |
Water |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Cofra Women's Safety Shoes, Paula Series > Size: 3