Learn More
WP Uranium, 15mL, SPEX CertiPrep™ Product Code.: 15216356
WP Uranium, 15mL, SPEX CertiPrep™
Product Code. 15216356 Shop All SPEX Certiprep Products
15 mL
Click to view available options
Quantity:
15 mL
Product Code. 15216356
Brand: SPEX Certiprep 4402
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Quantity 15 mL
Product Content Correction
Product Title
WP Uranium, 15mL, SPEX CertiPrep™ >