Learn More
uvex X-Fit Disposable Earplugs
X-Fit Disposable Earplugs with maximum insulation and optimum comfort.
Brand: uvex 2112022
55.60 EUR valid until 2025-06-08
Use promo code "24131" to get your promotional price.
This item is not returnable.
View return policy
Description
- The ergonomically pre-shaped uvex x-fit disposable earplugs provide very strong insulation and are suitable for use in extremely loud environments.
- The soft foam of the earplugs provides a high level of comfort, even when worn for extended periods.
Specifications
300 Pairs, Refill Box | |
PU Foam | |
Lime | |
Yes | |
Lime | |
Uncorded | |
Ear Plugs | |
No | |
Excellent speech and signal recognition |
EN 352-2 standard | |
Class AL | |
Regular | |
PU Foam | |
37 db | |
Oval | |
Single-use | |
Yes | |
EN 352-2 |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
Product Content Correction
Product Title
uvex X-Fit Disposable Earplugs > 300 Pairs, Refill Box