missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand X1000 UltraClean : 9mm PP Short Thread Cap,blue,,Silic. white/PTFE red,55° A,1.0mm,slitted

Brand:  Fisherbrand 09 15 1474

152.46 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


X1000 UltraClean : 9mm PP Short Thread Cap,blue,,Silic. white/PTFE red,55° A,1.0mm,slitted

Product Code. 15252805

  • 239.00€ / Pack of 1000
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand X1000 UltraClean : 9mm PP Short Thread Cap,blue,,Silic. white/PTFE red,55° A,1.0mm,slitted >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.