Learn More
Xanthine, 99%, Thermo Scientific Chemicals
76.98€ - 254.27€
Chemical Identifiers
CAS | 69-89-6 |
---|---|
Molecular Formula | C5H4N4O2 |
Molecular Weight (g/mol) | 152.113 |
MDL Number | MFCD00078453 |
InChI Key | LRFVTYWOQMYALW-UHFFFAOYSA-N |
Synonym | xanthine, 2,6-dihydroxypurine, xanthin, 2,6-dioxopurine, pseudoxanthine, 1h-purine-2,6 3h,7h-dione, isoxanthine, xanthic oxide, 1h-purine-2,6-diol, 9h-purine-2,6-diol |
PubChem CID | 1188 |
ChEBI | CHEBI:17712 |
IUPAC Name | 3,7-dihydropurine-2,6-dione |
SMILES | C1=NC2=C(N1)C(=O)NC(=O)N2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11488933
|
Thermo Scientific Alfa Aesar
A11077.14 |
25 g |
76.98€
25g |
In Stock
Log in to see stock. |
||||||
11498933
|
Thermo Scientific Alfa Aesar
A11077.22 |
100 g |
254.27€
100g |
In Stock
Log in to see stock. |
||||||
Description
Xanthine and xanthine oxidase system can be used to produce superoxide radicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsXanthine and xanthine oxidase system can be used to produce superoxide radicals.
Solubility
Soluble in water(0.067g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
69-89-6 | |
152.113 | |
LRFVTYWOQMYALW-UHFFFAOYSA-N | |
1188 | |
3,7-dihydropurine-2,6-dione |
C5H4N4O2 | |
MFCD00078453 | |
xanthine, 2,6-dihydroxypurine, xanthin, 2,6-dioxopurine, pseudoxanthine, 1h-purine-2,6 3h,7h-dione, isoxanthine, xanthic oxide, 1h-purine-2,6-diol, 9h-purine-2,6-diol | |
CHEBI:17712 | |
C1=NC2=C(N1)C(=O)NC(=O)N2 |
Specifications
69-89-6 | |
Odorless | |
MFCD00078453 | |
14,10059 | |
Soluble in water(0.067g/L). | |
C1=NC2=C(N1)C(=O)NC(=O)N2 | |
152.113 | |
CHEBI:17712 | |
99% |
>300°C | |
C5H4N4O2 | |
8733 | |
xanthine, 2,6-dihydroxypurine, xanthin, 2,6-dioxopurine, pseudoxanthine, 1h-purine-2,6 3h,7h-dione, isoxanthine, xanthic oxide, 1h-purine-2,6-diol, 9h-purine-2,6-diol | |
LRFVTYWOQMYALW-UHFFFAOYSA-N | |
3,7-dihydropurine-2,6-dione | |
1188 | |
152.11 | |
Xanthine |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 200-718-6
missing translation for 'rtecsNumber' : ZD7700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Xanthine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Xanthine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.