Learn More
Thermo Scientific Chemicals Yeast Nitrogen Base, without amino acids, without ammonium sulfate
Used for classifying yeasts based on carbohydrate and amino acids requirements. Recommended for use in classifying yeasts based on carbon and nitrogen requirements. Contains all essential nutrients required for the growth of yeasts except amino acids, nitrogen and carbohydrate.
213.06€ - 701.49€
Specifications
Form Powder
Packaging Type Plastic Bottle
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11336759
View Documents
Thermo Scientific Chemicals
H26271.36
500 g
701.49€
500g
11326759
View Documents
Thermo Scientific Chemicals
H26271.22
100 g
213.06€
100g
Description
Description
Yeast Nitrogen Base without Amino Acids and Ammonium Sulfate is used for classifying yeasts based on carbohydrate and amino acids requirements. Recommended for use in classifying yeasts based on carbon and nitrogen requirements. Contains all essential nutrients required for the growth of yeasts except amino acids, nitrogen and carbohydrate. Addition of nitrogen and carbon sources is required.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Powder
Plastic Bottle
Videos
Safety and Handling
Safety and Handling
Recommended Storage : Ambient temperatures
SDS
Documents
Documents
Product Certifications
Product Title
Thermo Scientific Chemicals Yeast Nitrogen Base, without amino acids, without ammonium sulfate