Learn More
Yeast-Torula, Thermo Scientific Chemicals
For biological research purposes.
389.00€ - 980.00€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15815208
|
Thermo Scientific
J23551.Q5 |
100 lb. |
980.00€
45359.20g |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||||
15805208
|
Thermo Scientific
J23551.Q1 |
25 lb. |
389.00€
11339.80g |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||||
Description
- Format: Solid
-
Specifications
Yeast-Torula | |
Solid |
RUO – Research Use Only