Learn More
YSI™ 2003 Pro Series Polarographic DO Sensor
Use with the Professional Plus, Pro20 or Pro2030 instruments
Brand:  YSI™ 605203
Product Code. 11967117
  • 203.00€ / Each
Estimated Shipment: 20-11-2024
Description
Use with the Professional Plus, Pro20 or Pro2030 instruments
  • Comes with 6 membrane caps and bottle of solution
  • Can be used on 60520 (DO), 6052030 (DO/conductivity), 6051020 (DO/ISE), and 605790 Quatro (DO/conductivity/ISE/ISE) cables
  • Easy user-replaceble sensor allows for quick sensor replacements; no tools required
  • Uses convenient screw-on cap membranes making membrane changes simple and easy
  • Order cables separately
Services
One year.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Professional Plus multiparameter meter and Pro20 DO meter and can be used on 60520 (DO), 6052030 (DO/conductivity), 6051020 (DO/ISE), 605790 Quatro (DO/conductivity/ISE/ISE) cables
1 Year
Polarographic DO Sensor
YSI 2003 DO module, 5908 cap membrane kit, instruction sheet, hex wrench, set screw