Learn More
YSI™ Probe Reconditioning Kit
For YSI Dissolved Oxygen and BOD Probes
Brand:  YSI™ 060745
Product Code. 12764566
  • 44.30€ / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Includes: Sanding tool and 10 discs
Description
Description
  • Reconditions 13-299-43 (Model 5739 Field Oxygen/Temperature Probe), 15-176-50 (Model 5718 Dissolved Oxygen Probe), 13-298-80 (Model 5750 BOD Probe), and 13-298-56, -57, and -62 (Model 55 Handheld Dissolved Oxygen Probes)
  • Removes tarnish and silver deposits from sensor's gold electrode
  • Maintains spherical contour of probe face
TRUSTED_SUSTAINABILITY
Specifications
Specifications
YSI™ 5718, 5739, 5750, 55 DO Probe
Reconditioning Kit
Sanding tool and 10 discs
DO