Learn More
Novus Biologicals™ Z-LE(OMe)HD(OMe)-FMK (Caspase 9 Inhibitor)
For use in research applications
484.00€ - 1370.00€
Specifications
Color | Tan |
---|---|
Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780484
|
Novus Biologicals™
NBP2-29398 |
484.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
18118672
|
Novus Biologicals™
NBP2-29398-5MG |
5 mg |
1370.00€
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
Tan | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles |
For Research Use Only
Product Title
Novus Biologicals™ Z-LE(OMe)HD(OMe)-FMK (Caspase 9 Inhibitor)