Learn More
Zeochem™ AG ZEOprep™ Classic
ZEOprep™ is a proven, highly successful packing material providing fast, effective and reproducible separations. Irregular shaped silica gel is used in Gravity, Flash and Medium Pressure Chromatography. Available in a variety of pore and particle sizes, moisture contents plus functionalities.
76.00€ - 5000.00€
Specifications
Technique | Chromatography |
---|---|
Type | Silica Gel |
Material | Silica Gel |
Particle Shape | Irregular |
Product Code | Brand | Pore Size | Particle Size | Phase | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Particle Size | Phase | Quantity | Price | Quantity & Availability | ||
30048966
|
Zeochem™ AG
301989 |
60 Å | 25 to 40 μm | Bare Silica | 1 kg |
101.00€
1kg |
In Stock
Log in to see stock. |
||
30048948
|
Zeochem™ AG
301988 |
60 Å | 25 to 40 μm | Bare Silica | 5 kg |
370.00€
5kg |
In Stock
Log in to see stock. |
||
30049033
|
Zeochem™ AG
300231 |
60 Å | 40 to 63 μm | Bare Silica | 1 kg |
85.00€
1kg |
In Stock
Log in to see stock. |
||
30048995
|
Zeochem™ AG
302097 |
60 Å | 40 to 63 μm | Bare Silica | 5 kg |
315.00€
5kg |
In Stock
Log in to see stock. |
||
30048957
|
Zeochem™ AG
300190 |
60 Å | 40 to 63 μm | C18 | 100 g |
249.00€
100g |
In Stock
Log in to see stock. |
||
30048967
|
Zeochem™ AG
302074 |
60 Å | 40 to 63 μm | C18 | 500 g |
867.00€
500g |
In Stock
Log in to see stock. |
||
30049024
|
Zeochem™ AG
300191 |
60 Å | 40 to 63 μm | C18 | 1 kg |
1335.00€
1kg |
In Stock
Log in to see stock. |
||
30049031
|
Zeochem™ AG
302089 |
60 Å | 40 to 63 μm | C18 | 5 kg |
4895.00€
5kg |
In Stock
Log in to see stock. |
||
30048965
|
Zeochem™ AG
300186 |
60 Å | 40 to 63 μm | Diol | 100 g |
249.00€
100g |
In Stock
Log in to see stock. |
||
30048949
|
Zeochem™ AG
302076 |
60 Å | 40 to 63 μm | Diol | 500 g |
911.00€
500g |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30049026
|
Zeochem™ AG
300187 |
60 Å | 40 to 63 μm | Diol | 1 kg |
1400.00€
1kg |
In Stock
Log in to see stock. |
||
30049002
|
Zeochem™ AG
302091 |
60 Å | 40 to 63 μm | Diol | 5 kg |
4770.00€
5kg |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30048962
|
Zeochem™ AG
300184 |
60 Å | 40 to 63 μm | NH2 | 100 g |
249.00€
100g |
In Stock
Log in to see stock. |
||
30048955
|
Zeochem™ AG
302080 |
60 Å | 40 to 63 μm | NH2 | 500 g |
911.00€
500g |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30048997
|
Zeochem™ AG
300185 |
60 Å | 40 to 63 μm | NH2 | 1 kg |
1400.00€
1kg |
In Stock
Log in to see stock. |
||
30048973
|
Zeochem™ AG
302092 |
60 Å | 40 to 63 μm | NH2 | 5 kg |
4755.00€
5kg |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30049021
|
Zeochem™ AG
300166 |
60 Å | 40 to 63 μm | SAX | 100 g |
249.00€
100g |
In Stock
Log in to see stock. |
||
30048988
|
Zeochem™ AG
302082 |
60 Å | 40 to 63 μm | SAX | 500 g |
938.00€
500g |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30049015
|
Zeochem™ AG
300167 |
60 Å | 40 to 63 μm | SAX | 1 kg |
1445.00€
1kg |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30049007
|
Zeochem™ AG
302094 |
60 Å | 40 to 63 μm | SAX | 5 kg |
4915.00€
5kg |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30048983
|
Zeochem™ AG
300177 |
60 Å | 40 to 63 μm | SH (Thiol) | 100 g |
249.00€
100g |
In Stock
Log in to see stock. |
||
30049005
|
Zeochem™ AG
302083 |
60 Å | 40 to 63 μm | SH (Thiol) | 500 g |
938.00€
500g |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30049030
|
Zeochem™ AG
300178 |
60 Å | 40 to 63 μm | SH (Thiol) | 1 kg |
1445.00€
1kg |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30049022
|
Zeochem™ AG
302095 |
60 Å | 40 to 63 μm | SH (Thiol) | 5 kg |
4915.00€
5kg |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30048991
|
Zeochem™ AG
300182 |
60 Å | 40 to 63 μm | NH2P | 100 g |
249.00€
100g |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30048984
|
Zeochem™ AG
302081 |
60 Å | 40 to 63 μm | NH2P | 500 g |
911.00€
500g |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30049020
|
Zeochem™ AG
300183 |
60 Å | 40 to 63 μm | NH2P | 1 kg |
1400.00€
1kg |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30048963
|
Zeochem™ AG
302093 |
60 Å | 40 to 63 μm | NH2P | 5 kg |
4770.00€
5kg |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30048958
|
Zeochem™ AG
300227 |
60 Å | 60 to 200 μm | Bare Silica | 1 kg |
76.00€
1kg |
In Stock
Log in to see stock. |
||
30048987
|
Zeochem™ AG
302098 |
60 Å | 60 to 200 μm | Bare Silica | 5 kg |
284.00€
5kg |
In Stock
Log in to see stock. |
||
30048975
|
Zeochem™ AG
300221 |
60 Å | 200 to 500 μm | Bare Silica | 1 kg |
76.00€
1kg |
In Stock
Log in to see stock. |
||
30048970
|
Zeochem™ AG
302205 |
60 Å | 200 to 500 μm | Bare Silica | 5 kg |
370.00€
5kg |
In Stock
Log in to see stock. |
||
30048952
|
Zeochem™ AG
302106 |
90 Å | 40 to 63 μm | Bare Silica | 5 kg |
354.00€
5kg |
In Stock
Log in to see stock. |
||
30048971
|
Zeochem™ AG
300175 |
90 Å | 40 to 63 μm | C18 | 100 g |
249.00€
100g |
In Stock
Log in to see stock. |
||
30049006
|
Zeochem™ AG
302075 |
90 Å | 40 to 63 μm | C18 | 500 g |
955.00€
500g |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30049017
|
Zeochem™ AG
300176 |
90 Å | 40 to 63 μm | C18 | 1 kg |
1470.00€
1kg |
In Stock
Log in to see stock. |
||
30048950
|
Zeochem™ AG
302090 |
90 Å | 40 to 63 μm | C18 | 5 kg |
5000.00€
5kg |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||
30049009
|
Zeochem™ AG
302107 |
90 Å | 60 to 200 μm | Bare Silica | 1 kg |
85.00€
1kg |
In Stock
Log in to see stock. |
||
30048976
|
Zeochem™ AG
302108 |
90 Å | 60 to 200 μm | Bare Silica | 5 kg |
327.00€
5kg |
In Stock
Log in to see stock. |
||
Description
- Irregular Amorphous Silica Gel
- High Purity
- European Manufactured
- Swiss accuracy and consistency
- Excellent batch to batch reproducibility
- ISO Certification
- Very wide portfolio
- Wide Selection of functionalization options
- Available with 4 different moisture contents
- High Chemical and mechanical stability
- Narrow Particle Size Distribution
- 20 μum to 500 μum particle sizes available
- Available in 60 A and 90 A pores sizes
- High loadability
- Unsurpassed Service and Support
- Selection of pack sizes available
- Suitable for gravity, flash and process
Specifications
Chromatography | |
Silica Gel | |
Silica Gel | |
Irregular |