missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™
34.60€ - 102.00€
Chemical Identifiers
CAS | 657-06-7 |
---|---|
Molecular Formula | C8H4ClF3O2 |
Molecular Weight (g/mol) | 224.563 |
MDL Number | MFCD00061094 |
InChI Key | WLXRKCGYQAKHSJ-UHFFFAOYSA-N |
Synonym | 2-chloro-5-trifluoromethyl benzoic acid, benzoic acid, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl benzoicacid, 2-chloro-5-trifluoromethyl-benzoic acid, maybridge1_003457, pubchem1377, pubchem7879, acmc-209xwb, 2-chloro5-trifluoromethylbenzoic acid |
PubChem CID | 688182 |
IUPAC Name | 2-chloro-5-(trifluoromethyl)benzoic acid |
SMILES | C1=CC(=C(C=C1C(F)(F)F)C(=O)O)Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
657-06-7 | |
224.563 | |
WLXRKCGYQAKHSJ-UHFFFAOYSA-N | |
688182 | |
C1=CC(=C(C=C1C(F)(F)F)C(=O)O)Cl |
C8H4ClF3O2 | |
MFCD00061094 | |
2-chloro-5-trifluoromethyl benzoic acid, benzoic acid, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl benzoicacid, 2-chloro-5-trifluoromethyl-benzoic acid, maybridge1_003457, pubchem1377, pubchem7879, acmc-209xwb, 2-chloro5-trifluoromethylbenzoic acid | |
2-chloro-5-(trifluoromethyl)benzoic acid |
Specifications
657-06-7 | |
C8H4ClF3O2 | |
2-chloro-5-trifluoromethyl benzoic acid, benzoic acid, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl benzoicacid, 2-chloro-5-trifluoromethyl-benzoic acid, maybridge1_003457, pubchem1377, pubchem7879, acmc-209xwb, 2-chloro5-trifluoromethylbenzoic acid | |
C1=CC(=C(C=C1C(F)(F)F)C(=O)O)Cl | |
224.563 | |
224.57 | |
2-Chloro-5-(trifluoromethyl)benzoic acid |
93°C to 96°C | |
MFCD00061094 | |
WLXRKCGYQAKHSJ-UHFFFAOYSA-N | |
2-chloro-5-(trifluoromethyl)benzoic acid | |
688182 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures