Learn More
3-Bromobenzhydrazide, 98+%, Thermo Scientific Chemicals
Description
3-Bromobenzhydrazide is used to produce 5-(3-bromo-phenyl)-3H-[1,3,4]oxadiazole-2-thione. This reaction will need reagent potassium hydroxide and solvent ethanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Bromobenzhydrazide is used to produce 5-(3-bromo-phenyl)-3H-[1,3,4]oxadiazole-2-thione. This reaction will need reagent potassium hydroxide and solvent ethanol.
Solubility
Slightly soluble in water (3.7 g/L) (25°C)
Notes
Keep container tightly closed. Store away from oxidizing agents.

Specifications
Specifications
Melting Point | 154°C to 157°C |
Quantity | 5 g |
Beilstein | 1945814 |
Solubility Information | Slightly soluble in water (3.7g/L) (25°C) |
Formula Weight | 215.05 |
Percent Purity | ≥98% |
Chemical Name or Material | 3-Bromobenzhydrazide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.