Learn More
3M™ Versaflo™ Air-Flow Adjustment Cover for Premium Head Suspension
Cover is used to adjust the location of the airflow in the hood by attaching to the end of the breathing tube. For use with S-950 Premium Head Suspension.
Brand: 3M™ 7000000157
9.05 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with S-950 Premium Head Suspension
- Attached to end of breathing tube, ring allows user to adjust location of airflow in hood
- 3M Catalog# S-955
Specifications
OSHA | |
Airflow Adjustment Cover | |
S-955 |
Reusable | |
Premium Head Suspension S-950 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
3M™ Versaflo™ Air-Flow Adjustment Cover for Premium Head Suspension > Air-Flow adjustment cover