Learn More
Linker Industrietechnik™ 50 Dia. x 25 mm Height Stainless Steel Test Sieves
Brand: Linker Industrietechnik™ 552.025.222-019
This item is not returnable.
View return policy
Specifications
1Each | |
Test Sieve | |
Stainless Steel | |
50 mm |
50 μm | |
Compliance 2.1 EN 10 204 and ISO 3310-1 | |
25mm | |
Stainless Steel |