missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Polystyrene Flat Bottom Strip Plates, 2 x 8 Strips
Suitable for optical measurements. Greiner Bio-One 96 Well Polystyrene Strip Plates, 2 x 8 Strips, F-Bottom are suitable for non polar proteins and peptides.
Brand: Greiner Bio-One 756071
This item is not returnable.
View return policy
Description
- Free of detectable DNase, RNase, human DNA
- 2 x 8 strips
- Non-Pyrogenic
- Light beam not deflected by well profile Suitable for transmission measurements
- Polystyrene surface is more hydrophobic than surface of hi
Specifications
Strip Plate | |
White | |
Polystyrene | |
20 to 350 μL | |
Microlon 600 | |
Flat Bottom (F-well) | |
100 | |
Yes | |
Clear |
High Binding | |
Strip Plate | |
6 | |
Non-sterile | |
388 μL | |
Yes | |
5 | |
-20°C to +60°C |