missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well Polystyrene Flat Bottom Strip Plates, 2 x 8 Strips

Suitable for optical measurements. Greiner Bio-One 96 Well Polystyrene Strip Plates, 2 x 8 Strips, F-Bottom are suitable for non polar proteins and peptides.

769.00€ - 831.00€

Specifications

Format Strip Plate
Color White
Material Polystyrene
No. of Wells 6
Volume (Metric) Working 20 to 350 μL
View More Specs

Products 2
Product Code Brand Surface Treatment Price Quantity & Availability  
Product Code Brand Surface Treatment Price Quantity & Availability  
10705982
View Documents Promotion Details
Greiner Bio-One
756070
Microlon 200
769.00€
Pack of 100
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
 
10690932
View Documents Promotion Details
Greiner Bio-One
756071
Microlon 600
831.00€
Pack of 100
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
 
Description

Description

  • Free of detectable DNase, RNase, human DNA
  • 2 x 8 strips
  • Non-Pyrogenic
  • Light beam not deflected by well profile
  • Suitable for transmission measurements
  • Polystyrene surface is more hydrophobic than surface of hi
Specifications

Specifications

Strip Plate
Polystyrene
20 to 350 μL
388 μL
Yes
5
-20°C to +60°C
White
6
Non-sterile
Flat Bottom (F-well)
100
Yes
Clear
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4 Years after month of production

SDS
Documents

Documents

Product Certifications