missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well Polystyrene Flat Bottom Strip Plates, 2 x 8 Strips

Suitable for optical measurements. Greiner Bio-One 96 Well Polystyrene Strip Plates, 2 x 8 Strips, F-Bottom are suitable for non polar proteins and peptides.

Brand:  Greiner Bio-One 756070

 View more versions of this product

Product Code. 10705982

  • 769.00€ / Pack of 100
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Free of detectable DNase, RNase, human DNA
  • 2 x 8 strips
  • Non-Pyrogenic
  • Light beam not deflected by well profile
  • Suitable for transmission measurements
  • Polystyrene surface is more hydrophobic than surface of hi
    TRUSTED_SUSTAINABILITY
Specifications

Specifications

Strip Plate
White
6
Non-sterile
388 μL
Yes
5
-20°C to +60°C
Medium Binding
Polystyrene
20 to 350 μL
Microlon 200
Flat Bottom (F-well)
100
Yes
Clear
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers