missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Polystyrene Round Bottom Strip Plates, 1 x 8 strips
Suitable for agglutination tests. Greiner Bio-One 96 Well Polystyrene Round Bottom Strip Plates, 1 x 8 strips are suitable for non polar proteins and peptides.
Brand: Greiner Bio-One 767070
This item is not returnable.
View return policy
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Pipetted simply and cleanly
- Suitable for +/-analyses
Specifications
Strip Plate | |
Medium Binding | |
Strip Plate | |
96 | |
Non-sterile | |
388 μL | |
Yes | |
Yes | |
Clear |
8 x 12 | |
White | |
Polystyrene | |
20 to 350 μL | |
Microlon 200 | |
Round Bottom (U-well) | |
100 | |
-20°C to +60°C |