missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well Polystyrene Round Bottom Strip Plates, 1 x 8 strips

Suitable for agglutination tests. Greiner Bio-One 96 Well Polystyrene Round Bottom Strip Plates, 1 x 8 strips are suitable for non polar proteins and peptides.

1055.00€ - 1225.00€

Specifications

Format Strip Plate
Array 8 x 12
Color White
Material Polystyrene
No. of Wells 96
View More Specs

Products 2
Product Code Brand Surface Treatment Price Quantity & Availability  
Product Code Brand Surface Treatment Price Quantity & Availability  
10329181
View Documents Promotion Details
Greiner Bio-One
767070
Microlon 200
1055.00€
Pack of 100
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
 
10207211
View Documents Promotion Details
Greiner Bio-One
767071
Microlon 600
1225.00€
Pack of 100
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
 
Description

Description

  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Pipetted simply and cleanly
  • Suitable for +/-analyses
Specifications

Specifications

Strip Plate
White
96
Non-sterile
Round Bottom (U-well)
100
-20°C to +60°C
8 x 12
Polystyrene
20 to 350 μL
388 μL
Yes
Yes
Clear
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4 Years after month of production

SDS
Documents

Documents

Product Certifications