Learn More
Greiner Bio-One 96-well Polystyrene Round Bottom Strip Plates, 2 x 8 Strips
Treated specifically to increase protein binding. Greiner Bio-One 96 Well Polystyrene Round Bottom Strip Plates, 2 x 8 Strips are suited for agglutination tests.
820.00€ - 861.00€
Specifications
Format | Strip Plate |
---|---|
Color | White |
Material | Polystyrene |
No. of Wells | 6 |
Volume (Metric) Working | 50 to 280 μL |
Product Code | Brand | Surface Treatment | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Surface Treatment | Price | Quantity & Availability | |||||
10778242
|
Greiner Bio-One
754070 |
Microlon 200 |
861.00€
Pack of 100 |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||||
10660552
|
Greiner Bio-One
754061 |
Microlon 600 |
820.00€
Pack of 100 |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||||
Description
- Free of detectable DNase, RNase, human DNA
- 2 x 8 strips
- Non-Pyrogenic
- Pipetted simply and cleanly
- Suitable for +/- analyses
- Suitable for transmission measurements
- U-bottom (round bottom)
Specifications
Strip Plate | |
Polystyrene | |
50 to 280 μL | |
312 μL | |
Yes | |
5 | |
-20°C to +60°C |
White | |
6 | |
Non-sterile | |
Round Bottom (U-well) | |
100 | |
Yes | |
Clear |
Safety and Handling
missing translation for 'shelfLife' : 4 Years after month of production