missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Abpure™ TCS Antibody Purification Kit
Novus Biologicals™ Abpure TCS Antibody Purification Kit can be used in its rapid version by spin-purification in centrifuge, or by gravity-flow purification.
504.00€ - 1115.00€
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
18057074
|
Novus Biologicals™
264-0500 |
3 Purifications |
1115.00€
3 tests |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
18066404
|
Novus Biologicals™
264-0030 |
1 Purification |
504.00€
1 test |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
Description
- Fully compatible with InnovaCoat GOLD and LATEX conjugation kits which allow purified antibody to be conjugated rapidly
- Antibody to be purified should be in 10mL to 50mL of tissue culture supernatant (TCS)
- Up to 5mg of antibody can be purified in each run
Specifications
4°C | |
Antibody Purification Kit |
For research use only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title