Learn More
IKA™ Accessory Stirring Shafts for SteadyStir™ and Overhead Stirrers
Generate axial flow in the vessel with these stirring shafts. IKA™ Accessory Stirring Shafts for SteadyStir™ and IKA™ Overhead Stirrers are designed for medium to high speeds.
Brand: IKA™ 0000756900
183.47 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- For stirring in round vessels with narrow necks
- Effect is similar to that of a 4-bladed propeller stirrer
- Medium to high speeds required
† PTFE-coated
Specifications
0.3 in. | |
13.8 in. | |
R 1352 | |
Centrifugal Stirrer |
8 mm | |
350 mm | |
2000 rpm max. | |
Stainless Steel (316L) |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30