Learn More
Ansell AlphaTec™ 55-100 NRL RABS and Isolator Gloves Product Code.: 16885230
Add to basket
Ansell AlphaTec™ 55-100 NRL RABS and Isolator Gloves
Product Code. 16885230 Shop All Ansell Products
8
Click to view available options
Size:
8
9
10
Product Code. 16885230
Brand: Ansell 55100080
Add to basket
Add to basket
Gloves provide effective protection for medium duty physical environments, especially for defense against abrasions and tears. Constructed of 0.51 mm latex to ensure maximum dexterity in crucial applications.
  • Latex construction provides defense against tears, abrasions, cuts, and punctures
  • Ideal protection for medium duty physical environments
  • Translucent and unlined for effective cleaning
  • A variety of port sizes are available to fit most isolator glove boxes
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009
Content And Storage When not in use store in a dark, dry environment in original packaging
Color Natural
Hand Specific
Latex Free No
Material Latex
Size 8
Thickness (English) 20 mil
Thickness (Metric) 0.51 mm
EN388 Standard 2020X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES KLNOPT
For Use With (Application) Medium Duty Environments
For Use With (Equipment) Isolator Glove Boxes
Port Size (English) 6 in.
Port Size (Metric) 152 mm
Series 55-100
Show More Show Less
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
Ansell AlphaTec™ 55-100 NRL RABS and Isolator Gloves >