missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Antibody Array Detection Kit

Antibody array detection kit is designed to enable each antibody microarray.

Brand:  Abnova™ AA0135.1Kit


Product Code. 16002545

  • 738.00€ / 1 set
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For best results, store the components as described in the protocol
  • Avoid repeated freezing and thawing
Specifications

Specifications

Store at -20°C. Avoid freeze-thaw cycles.
1 Kit
Profiling
Array/Library
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Antibody Array Detection Kit

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Supplemental information
  • MIXTURE LIST-Contains : Triton X100, Tween 20

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.