missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ APEG1 Recombinant Protein

Brand:  Abnova™ H00010290-P01.25ug

 View more versions of this product

Product Code. 16104842

  • 521.00€ / 25µg
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH06346
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
38.17
8
Glutathione Sepharose 4 Fast Flow
25 μg
MKPSPSQNRRSSDTGSKAPPTFKVSLMDQSVREGQDVIMSIRVQGEPKPVVSWLRNRQPVRPDQRRFAEEAEGGLCRLRILAAERGDAGFYTCKAVNEYGARQCEARLEVRGE
APEG1/BPEG/KIAA1297/MGC12676/SPEGalpha/SPEGbeta
SPEG
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
10290
APEG1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SPEG
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.