Learn More
Thermo Scientific Chemicals Apelin-13, Human, Bovine
For cellular and molecular biology applications
Brand:  Thermo Scientific Chemicals J66620.|D
 View more versions of this product
Product Code. 15468049
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
An endogenous ligand of Apelin Receptor (APLNR/APJ receptor) that plays important roles in regulating fluid homeostasis, cardiovascular function, and insulin sensitivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human, Bovine
-30°C to -10°C
1550.82
0.5 mg
Gln-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe
HPLC
Lyophilized Powder
Apelin-13, human, bovine
RUO - research use only