missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Apelin-13, Human, Bovine

For cellular and molecular biology applications

101.00€ - 118.59€

Specifications

Species Human, Bovine
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Lyophilized Powder
Molecular Weight (g/mol) 1550.82
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15468049
View Documents
Thermo Scientific Chemicals
J66620.|D
0.5 mg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
15478049
View Documents
Thermo Scientific Chemicals
J66620.M^
1 mg
118.59€
1mg
In Stock
to see stock.
Add to basket
 
Description

Description

An endogenous ligand of Apelin Receptor (APLNR/APJ receptor) that plays important roles in regulating fluid homeostasis, cardiovascular function, and insulin sensitivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Human, Bovine
-30°C to -10°C
1550.82
RUO - research use only
HPLC
Lyophilized Powder
Apelin-13, human, bovine
Gln-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only