missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ARF7 Recombinant Protein

344.00€ - 521.00€

Specifications

Accession Number AAH34354
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 80117
Molecular Weight (g/mol) 46.86
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16194763
View Documents
Abnova™
H00080117-P01.10ug
10 μg
344.00€
10µg
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
16104773
View Documents
Abnova™
H00080117-P01.25ug
25 μg
521.00€
25µg
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH34354
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
46.86
8
Glutathione Sepharose 4 Fast Flow
Wheat Germ (in vitro)
ARF7/FLJ22595
ARL14
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
80117
FLJ22595 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARL14
Human
Recombinant
Solution
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.