Learn More
Deltalab™ Autoclave Resistant Bags
Wrap autoclaveable items in Deltalab™Autoclave Resistant Bags when working with laboratory items requiring sterilization.
Brand:  Deltalab™ 200310
 View more versions of this product
Product Code. 12054363
  • 364.00€ / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Printed with biohazard graphic symbol and use instructions in five languages
  • ags placed in autoclave at 121ºC. should not be sealed hermetically
  • Minimum order quantity: 50 pieces
Specifications
Specifications
Autoclavable
500 Pack
30 x 60cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Deltalab™ Autoclave Resistant Bags > Dimensions (L x W): 30 x 60cm