Learn More
Deltalab™ Autoclave Resistant Bags
Wrap autoclaveable items in Deltalab™Autoclave Resistant Bags when working with laboratory items requiring sterilization.
Brand: Deltalab™ 200311
Description
- Printed with biohazard graphic symbol and use instructions in five languages
- ags placed in autoclave at 121ºC. should not be sealed hermetically
- Minimum order quantity: 50 pieces
Specifications
Autoclavable | |
300 Pack | |
40 x 75cm |
Product Suggestions
Customers who viewed this item also viewed