Learn More
Thermo Scientific™ BactiSwab™ Gel Collection and Transport Swabs: Dual Swab Amies Clear
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
Brand:  Thermo Scientific™ R723000
 View more versions of this product
Product Code. 11992642
  • 1040.00€ / Pack of 500
Explore more special offers
Description
Description
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.
  • Blue cap
  • Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
  • Exclusive zip-seal packaging to promote optimum media performance
  • Color-coded cap with protective overskirt
  • Inert, non-toxic rayon swab tip for good sample retrieval and absorption
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Amies Clear
Blue
500/Pk.
Plastic
Single Applicator
Amies Clear
1 Tube/Envelope
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ BactiSwab™ Gel Collection and Transport Swabs: Dual Swab Amies Clear > Amies Clear; Sgl. plastic-shaft; Blue Cap; 500/Pk.