Learn More
Thermo Scientific™ BactiSwab™ Gel Collection and Transport Swabs: Dual Swab Amies Clear
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
1040.00€
Specifications
Media Type | Amies Clear |
---|---|
Closure Color | Blue |
Description | Amies Clear |
Quantity | 1 Tube/Envelope |
Shaft Material | Plastic |
Product Code | Brand | Packaging | Swab Configuration | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging | Swab Configuration | Price | Quantity & Availability | ||||
11992642
|
Thermo Scientific™
R723000 |
500/Pk. | Single Applicator |
1040.00€
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.- Blue cap
- Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
- Exclusive zip-seal packaging to promote optimum media performance
- Color-coded cap with protective overskirt
- Inert, non-toxic rayon swab tip for good sample retrieval and absorption
Specifications
Amies Clear | |
Amies Clear | |
Plastic |
Blue | |
1 Tube/Envelope |
Product Title
Thermo Scientific™ BactiSwab™ Gel Collection and Transport Swabs: Dual Swab Amies Clear