Learn More
Saint Gobain Life Sciences BarbLock™ Hand Tool
BarbLock™ Hand Tool with stainless steel jaw and handle installs BarbLock™ retainers to tubing.
Brand:  Saint Gobain Life Sciences BLT135187
 View more versions of this product
Product Code. 30040266
  • 285.50€ / Each
Estimated Shipment: 28-11-2024
Description
BarbLock™ ultra-secure retainers are the most advanced design ideal for systems transferring delicate liquids or gases. They have become the standard in many industries because they solve inherent leak path problems in sensitive and demanding applications. They require no adhesives or solvents and are available in high temperature materials for thermally demanding environments.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tubing Clamp
187 Series