Learn More
Saint Gobain Life Sciences BarbLock™ Hand Tool
BarbLock™ Hand Tool with stainless steel jaw and handle installs BarbLock™ retainers to tubing.
285.50€
Product Code | Brand | Product Line | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Line | Price | Quantity & Availability | |||||
30040264
|
Saint Gobain Life Sciences
BLT135125 |
125 Series |
285.50€
Each |
Estimated Shipment: 28-11-2024
Log in to see stock. |
|||||
30040266
|
Saint Gobain Life Sciences
BLT135187 |
187 Series |
285.50€
Each |
Estimated Shipment: 28-11-2024
Log in to see stock. |
|||||
30040253
|
Saint Gobain Life Sciences
BLT135250 |
250 Series |
285.50€
Each |
Estimated Shipment: 28-11-2024
Log in to see stock. |
|||||
30040257
|
Saint Gobain Life Sciences
BLT135252 |
252 Series |
285.50€
Each |
Estimated Shipment: 28-11-2024
Log in to see stock. |
|||||
30040236
|
Saint Gobain Life Sciences
BLT135500 |
500 Series |
285.50€
Each |
Estimated Shipment: 28-11-2024
Log in to see stock. |
|||||
Description
BarbLock™ ultra-secure retainers are the most advanced design ideal for systems transferring delicate liquids or gases. They have become the standard in many industries because they solve inherent leak path problems in sensitive and demanding applications. They require no adhesives or solvents and are available in high temperature materials for thermally demanding environments.
Specifications
Tubing Clamp |