Learn More
Saint Gobain Life Sciences BarbLock™ Removal Tool
Universal removal tool.
Brand:  Saint Gobain Life Sciences BLTURT
 View more versions of this product
Product Code. 30040239
  • 136.22€ / Each
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
BarbLock™ ultra-secure retainers are the most advanced design ideal for systems transferring delicate liquids or gases. They have become the standard in many industries because they solve inherent leak path problems in sensitive and demanding applications. They require no adhesives or solvents and are available in high temperature materials for thermally demanding environments.
  • Constructed from high-performance polypropylene that meets USP Class VI criteria
  • Quick and easy to assemble
  • Compatible with all flexible tubing material
  • Disposable assemblies
  • Eliminates solvent bonding, ultraviolet bonding and toxic adhesives
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tubing Clamp
500 Series