Learn More
Saint Gobain Life Sciences BarbLock™ Removal Tool
Universal removal tool.
136.22€ - 243.72€
Specifications
Product Type Tubing Clamp
View More Specs
Products 2
Product Code Brand Product Line Price Quantity & Availability  
Product Code Brand Product Line Price Quantity & Availability  
30040239
View Documents Promotion Details
Saint Gobain Life Sciences
BLTURT
500 Series
136.22€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-11-2024
to see stock.
Add to basket
30040267
View Documents Promotion Details
Saint Gobain Life Sciences
BLTURT II
625 Series
243.72€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-11-2024
Description
BarbLock™ ultra-secure retainers are the most advanced design ideal for systems transferring delicate liquids or gases. They have become the standard in many industries because they solve inherent leak path problems in sensitive and demanding applications. They require no adhesives or solvents and are available in high temperature materials for thermally demanding environments.
  • Constructed from high-performance polypropylene that meets USP Class VI criteria
  • Quick and easy to assemble
  • Compatible with all flexible tubing material
  • Disposable assemblies
  • Eliminates solvent bonding, ultraviolet bonding and toxic adhesives
  • Autoclavable
Specifications
Specifications
Tubing Clamp