Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Piperacillin/Tazobactam60
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
Brand:  BD 231692
Additional Details : Weight : 0.00870kg
 View more versions of this product
Product Code. 17362401
  • 166.00€ / Pack of 10
Estimated Shipment: 07-08-2024
Description
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Specifications
Piperacillin, Tazobactam 60
100/10μg
Disc
10 Pack
Special Offers
For in vitro diagnostic use