Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Piperacillin/Tazobactam60
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
166.00€
Specifications
Antimicrobial Agents | Piperacillin, Tazobactam 60 |
---|---|
Concentration | 100/10μg |
Packaging Type | Disc |
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Piperacillin, Tazobactam 60 | |
100/10μg | |
Disc |
For in vitro diagnostic use