missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD H-2Kb:Ig Recombinant Fusion Protein, Mouse
For immunoassay protocols
Brand: BD 550750
682.45 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
The H-2K[b]:Ig fusion protein consists of three extracellular major histocompatibility complex (MHC) class I H-2Kb domains that are fused to VH regions of mouse IgG1.- Format: PE
- Reactivity: Ms
- Isotype: Mouse IgG1, Λ

Specifications
0.5mg/mL | |
Aqueous Buffered solution containing 0.09% Sodium Azide | |
7.2 | |
0.25 mg | |
Recombinant |
ELISA, Flow Cytometry | |
Mouse H-2Kb:Ig Recombinant Fusion Protein | |
Affinity Chromatography, SDS-PAGE | |
4°C | |
Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD H-2Kb:Ig Recombinant Fusion Protein, Mouse > Recombinant Dimeric Mouse H-2K(b):Ig Fusion Protein; 0.25mg
Research Use Only
Spot an opportunity for improvement?Share a Content Correction