Learn More
BD Recombinant Human IL-12 (p70)
Potent regulator of cell-mediated immune responses
Brand:  BD 554613
352.54 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15857808
  • 353.00€ / Each
Explore more special offers
Description
Description
  • Supplied as a frozen liquid comprised of 0.22μm sterile-filtered aqueous buffered solution containing 1mg/mL Biotechnology grade, low endotoxin bovine serum albumin, with no preservatives
Enzyme Linked Immunosorbent Assay, BioAssay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100μg/mL
IL-12 (p70) Recombinant Protein
-80°C
Bioassay, ELISA Standard
5 μg
Recombinant
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Recombinant Human IL-12 (p70) > 5μg