Learn More
BD Recombinant Human IL-12 (p70)
Potent regulator of cell-mediated immune responses
Brand: BD 554613
352.54 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Supplied as a frozen liquid comprised of 0.22μm sterile-filtered aqueous buffered solution containing 1mg/mL Biotechnology grade, low endotoxin bovine serum albumin, with no preservatives
Enzyme Linked Immunosorbent Assay, BioAssay
Specifications
100μg/mL | |
IL-12 (p70) Recombinant Protein | |
-80°C |
Bioassay, ELISA Standard | |
5 μg | |
Recombinant |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
BD Recombinant Human IL-12 (p70) > 5μg