Learn More
Sartorius™ Biohit™ Proline™ Fixed Volume Pipettes
Single channel fixed volume pipettes.
Brand: Sartorius™ 722001
179.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Single channel fixed volume pipettes.
Specifications
1 | |
Fixed Volume Pipette | |
5 μL |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
38Hours
4Minutes
33Seconds
12New Offer Save Up to 92%
Product Content Correction
Product Title
Sartorius™ Biohit™ Proline™ Fixed Volume Pipettes >