Learn More
Fisherbrand™ Black Phenolic Caps
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
Quantity:
12
576
1600
3100
5500
Specifications
Specifications
Color | Black |
Quantity | 12 |
Closure Color | Black |
Neck Finish | 18-400 |
Closure Included | No |
Product Type | Lined Cap |
Closure Liner | Polyethylene Cone |
Closure Material | Phenolic |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Product Title
Fisherbrand™ Black Phenolic Caps >