Learn More
Brady™ BMP61 M611 Nylon Labels
Wrap-around labels are highly pliable to conform to tightly curved surfaces such as vials.
Brand:  Brady™ 150892
 View more versions of this product
Product Code. 17120441
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
  • Service temperature of -40 to 193° F (-40 to 90° C)
  • Low-profile material that adheres to surfaces that are smooth, rough or powder coated, as well as to irregular surfaces or surfaces with low surface energy
Specifications
Specifications
Acrylic
Nylon
Tape
BMP™61 Labeling System, BradyPrinter™ M611
0.0065 in.
30 ft.
White
Thermal Transfer
Laboratory ID - Bottle, Tube & Vial
1 in W x 30 ft L
30 ft. x 1 in.
1 in.