Learn More
Brady™ BMP71 BMP61 M611 Permanent Vinyl General Identification Labels
Permanent Vinyl (B-439) is a thermal transfer printable material ideal for applications needing various colors like finished product ID, rating plates and general purpose ID.
Brand:  Brady™ 011187
 View more versions of this product
Product Code. 11532545
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
  • Labels for the BMP61 and M611 Thermal Transfer Portable Printers
  • Can also be printed on the BMP™71 Label Printer provided the roll adapter (M71-ADAPT) is used
Specifications
Specifications
Permanent Acrylic
Gloss
Thermal Transfer
15.24 m x 12.7 mm
Black
Polyester (B-439)
BMP61, BMP71 (with Media Adapter), M611